accessibility menu, dialog, popup

UserName

CH3NCH3N

CAS RN 123-32-0

IUPAC Name: 2,5-dimethylpyrazine
Synonyms: 2,5-dimethylpyrazine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: LCZUOKDVTBMCMX-UHFFFAOYSA-N
SMILES: CC1=CN=C(C)C=N1
Boiling Point 
  • (3)
  • (5)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (3)
  • (5)
Physical Form 
  • (10)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (3)
  • (5)
  • (2)
  • (2)
  • (8)
  • (2)
  • (3)
  • (5)
  • (10)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)