missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine (contains 2,6-isomer) 80.0+%, TCI America™

Catalog No. p-7126768
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D152625ML 25 mL
D1526500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D152625ML Supplier TCI America Supplier No. D152625ML
Only null left

Chemical Identifiers

CAS 123-32-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006147
InChI Key LCZUOKDVTBMCMX-UHFFFAOYSA-N
Synonym 2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
PubChem CID 31252
IUPAC Name 2,5-dimethylpyrazine
SMILES CC1=CN=C(C=N1)C

Specifications

Color Yellow
Boiling Point 182°C
Quantity 25 mL
Formula Weight 108.14
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylpyrazine (contains 2,6-isomer)
EINECSNumber (5)-0950
RTECSNumber UQ2800000
TSCA Yes