missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D35171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35171G Supplier TCI America Supplier No. D35171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Organic compound is presented as a white to light yellow powder or crystal.

Chemical Identifiers

CAS 85199-06-0
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01863525
InChI Key OOMZKLJLVGQZGV-UHFFFAOYSA-N
Synonym 2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid, boronic acid, 2,5-dimethylphenyl, pubchem1831, p-xyleneboronic acid, ksc448a1r, 2,5-dimehylphenylboronic acid
PubChem CID 2734347
IUPAC Name (2,5-dimethylphenyl)boronic acid
SMILES CC1=CC=C(C)C(=C1)B(O)O

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 1 g
Packaging Glass Bottle
Physical Form Crystal-Powder at 20°C
Formula Weight 149.98
Percent Purity 97.0 to 114.0%
Grade Not Available
Chemical Name or Material 2,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride)