missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenyl isocyanate, 97%

CAS: 40397-98-6 | C9H9NO | 147.18 g/mol

$44.14 - $96.74

Chemical Identifiers

CAS 40397-98-6
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00013854
InChI Key SOXVXJQIQVOCAY-UHFFFAOYSA-N
Synonym 2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
PubChem CID 98605
ChEBI CHEBI:60097
IUPAC Name 2-isocyanato-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(=C1)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1599903
View Documents
Thermo Scientific Chemicals
L1599903
1 g
Each for $44.14
Only null left
 
AAL1599906
View Documents
Thermo Scientific Chemicals
L1599906
5 g
Each for $96.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40397-98-6
147.18
SOXVXJQIQVOCAY-UHFFFAOYSA-N
98605
2-isocyanato-1,4-dimethylbenzene
C9H9NO
MFCD00013854
2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
CHEBI:60097
CC1=CC=C(C)C(=C1)N=C=O

Specifications

40397-98-6
210°C
1.5315
MFCD00013854
1100841
SOXVXJQIQVOCAY-UHFFFAOYSA-N
2-isocyanato-1,4-dimethylbenzene
98605
147.18
2,5-Dimethylphenyl isocyanate
1.043
1 g
C9H9NO
UN2206
2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
CC1=CC=C(C)C(=C1)N=C=O
147.18
CHEBI:60097
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 000-000-0

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only