Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenyl isocyanate, 97%

Catalog No. AAL1599903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1599903 1 g
AAL1599906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1599903 Supplier Thermo Scientific Chemicals Supplier No. L1599903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40397-98-6 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40397-98-6
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00013854
InChI Key SOXVXJQIQVOCAY-UHFFFAOYSA-N
Synonym 2,5-dimethylphenyl isocyanate, 2,5-dimethylphenylisocyanate, benzene, 2-isocyanato-1,4-dimethyl, 2,5-dimethyl phenylisocyanate, 2,5-dimethylbenzenisocyanate, acmc-1art9, 1-isocyanato-2,5-dimethyl-benzene
PubChem CID 98605
ChEBI CHEBI:60097
IUPAC Name 2-isocyanato-1,4-dimethylbenzene
SMILES CC1=CC=C(C)C(=C1)N=C=O

Specifications

Density 1.043
Quantity 1 g
Refractive Index 1.5315
UN Number UN2206
Beilstein 1100841
Boiling Point 210°C
Formula Weight 147.18
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylphenyl isocyanate
Hazard Category H227-H302+H312+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P342+P311-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 000-000-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only