missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenol 98.0+%, TCI America™

Catalog No. p-7126424
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D077525G 25 g
D0775100G 100 g
D0775500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D077525G Supplier TCI America Supplier No. D077525G

Chemical Identifiers

CAS 95-87-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00002237
InChI Key NKTOLZVEWDHZMU-UHFFFAOYSA-N
Synonym 2,5-xylenol, p-xylenol, phenol, 2,5-dimethyl, 3,6-dimethylphenol, 6-methyl-m-cresol, 3,6-xylenol, 2,5-dimethyl phenol, 2,5-dmp, 1,2,5-xylenol, 1-hydroxy-2,5-dimethylbenzene
PubChem CID 7267
IUPAC Name 2,5-dimethylphenol
SMILES CC1=CC=C(C)C(O)=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
UN Number 2261
Boiling Point 212°C
Formula Weight 122.17
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,5-Dimethylphenol
EINECSNumber (3)-0521&(4)-0057
RTECSNumber ZE5775000
TSCA Yes