missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, 98+%

CAS: 625-86-5 | C6H8O | 96.129 g/mol

Supplier:  Thermo Scientific Chemicals A1283336

 View more versions of this product

Catalog No. AAA1283336


Only null left
Explore available promotions

Description

Description

2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

Solubility
Slightly miscible with water. Miscible with ethanol and fats.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications

Specifications

2,5-Dimethylfuran
-62°C
92°C to 93°C
C6H8O
MFCD00003250
UN3271
furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
GSNUFIFRDBKVIE-UHFFFAOYSA-N
2,5-dimethylfuran
12266
≥98%
625-86-5
0.903
1°C (33°F)
1.441
500 g
106449
Slightly miscible with water. Miscible with ethanol and fats.
CC1=CC=C(O1)C
96.129
96.13

Chemical Identifiers

625-86-5
96.129
GSNUFIFRDBKVIE-UHFFFAOYSA-N
12266
CC1=CC=C(O1)C
C6H8O
MFCD00003250
furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
2,5-dimethylfuran
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H315-H319-H317-H335
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c

H225-H302-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.

EINECSNumber : 210-914-3

RTECSNumber : LU0875000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only