missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylfuran, 98+%

Catalog No. p-7020769
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1283314 25 g
AAA1283322 100 g
AAA1283336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1283314 Supplier Thermo Scientific Chemicals Supplier No. A1283314

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 625-86-5 | C6H8O | 96.129 g/mol

2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylfuran is a major metabolite of hexane with singlet oxygen scavenging ability. It is used as biofuel, biomaker for smoking, pharmaceutical intermediate and in organic synthesis. It is also used to analyze samples for the derivatization of carbonyl-containing compounds to their oximes.

Solubility
Slightly miscible with water. Miscible with ethanol and fats.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 625-86-5
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00003250
InChI Key GSNUFIFRDBKVIE-UHFFFAOYSA-N
Synonym furan, 2,5-dimethyl, 2,5-dimethylfurane, 2,5-dimethyl-furan, 2,5-dimethyl furane, unii-dr5hl9oj7y, ccris 3158, 2,5-dimethyl furan, dr5hl9oj7y, 2.5-dimethyl furan, acmc-1avpg
PubChem CID 12266
IUPAC Name 2,5-dimethylfuran
SMILES CC1=CC=C(O1)C

Specifications

Melting Point -62°C
Density 0.903
Boiling Point 92°C to 93°C
Flash Point 1°C (33°F)
Refractive Index 1.441
Quantity 25 g
UN Number UN3271
Beilstein 106449
Solubility Information Slightly miscible with water. Miscible with ethanol and fats.
Formula Weight 96.13
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylfuran
Show More Show Less
Hazard Category H225-H302-H315-H317-H319-H335
Hazard Statement GHS H Statement

H225-H301-H315-H319-H317-H335

Highly flammable liquid and vapour.

Toxic if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause an allergic skin reaction.

May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 210-914-3
RTECSNumber LU0875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only