missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylcyclohexanone (mixture of isomers) 95.0+%, TCI America™

Catalog No. D12795ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D12795ML 5 mL
D127925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D12795ML Supplier TCI America Supplier No. D12795ML

Chemical Identifiers

CAS 932-51-4
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.20
MDL Number MFCD00045551
InChI Key LUGFNAYTNQMMMY-UHFFFAOYNA-N
PubChem CID 523055
IUPAC Name 2,5-dimethylcyclohexan-1-one
SMILES CC1CCC(C)C(=O)C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 mL
UN Number 1224
Formula Weight 126.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylcyclohexanone (mixture of isomers)
TSCA No
Recommended Storage Refrigerator