accessibility menu, dialog, popup

UserName

CH3OCH3

CAS RN 932-51-4

IUPAC Name: 2,5-dimethylcyclohexan-1-one
Synonyms: 2,5-dimethylcyclohexan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: LUGFNAYTNQMMMY-UHFFFAOYNA-N
SMILES: CC1CCC(C)C(=O)C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results