missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethylbenzyl Chloride 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 121°C |
| Quantity | 25 g |
| UN Number | 3265 |
| Formula Weight | 154.64 |
| Percent Purity | ≥98.0% (T,GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,5-Dimethylbenzyl Chloride |
Safety and Handling
| TSCA | Yes |