missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzyl chloride, 98%

CAS: 824-45-3 | C9H11Cl | 154.637 g/mol

$52.96 - $181.82

Chemical Identifiers

CAS 824-45-3
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00000902
InChI Key PECXPZGFZFGDRD-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzyl chloride, 2-chloromethyl-1,4-dimethylbenzene, 1-chloromethyl-2,5-dimethylbenzene, 2-chloromethyl-p-xylene, benzene, 2-chloromethyl-1,4-dimethyl, 2-chloromethyl-1,4-dimethyl-benzene, benzenemethanol,2-bromo-5-fluoro, 2,5-dimethylbenzylchloride, acmc-209poz, 2.5-dimethylbenzylchloride
PubChem CID 69987
IUPAC Name 2-(chloromethyl)-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)CCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0130306
View Documents
Thermo Scientific Chemicals
L0130306
5 g
Each for $52.96
Only null left
 
AAL0130314
View Documents
Thermo Scientific Chemicals
L0130314
25 g
Each for $181.82
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

824-45-3
154.637
PECXPZGFZFGDRD-UHFFFAOYSA-N
69987
CC1=CC(=C(C=C1)C)CCl
C9H11Cl
MFCD00000902
2,5-dimethylbenzyl chloride, 2-chloromethyl-1,4-dimethylbenzene, 1-chloromethyl-2,5-dimethylbenzene, 2-chloromethyl-p-xylene, benzene, 2-chloromethyl-1,4-dimethyl, 2-chloromethyl-1,4-dimethyl-benzene, benzenemethanol,2-bromo-5-fluoro, 2,5-dimethylbenzylchloride, acmc-209poz, 2.5-dimethylbenzylchloride
2-(chloromethyl)-1,4-dimethylbenzene

Specifications

824-45-3
232°C to 233°C
Pungent
1.537
5 g
2206277
PECXPZGFZFGDRD-UHFFFAOYSA-N
2-(chloromethyl)-1,4-dimethylbenzene
69987
98%
1.05
65°C (149°F)
C9H11Cl
MFCD00000902
UN3265
2,5-dimethylbenzyl chloride, 2-chloromethyl-1,4-dimethylbenzene, 1-chloromethyl-2,5-dimethylbenzene, 2-chloromethyl-p-xylene, benzene, 2-chloromethyl-1,4-dimethyl, 2-chloromethyl-1,4-dimethyl-benzene, benzenemethanol,2-bromo-5-fluoro, 2,5-dimethylbenzylchloride, acmc-209poz, 2.5-dimethylbenzylchloride
CC1=CC(=C(C=C1)C)CCl
154.637
154.64
2,5-Dimethylbenzyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 212-529-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only