missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzyl chloride, 98%

Numéro de catalogue. AAL0130314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL0130314 25 g
AAL0130306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL0130314 Fournisseur Thermo Scientific Chemicals Code fournisseur L0130314
Il en reste null

CAS: 824-45-3 | C9H11Cl | 154.637 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 824-45-3
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00000902
InChI Key PECXPZGFZFGDRD-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzyl chloride, 2-chloromethyl-1,4-dimethylbenzene, 1-chloromethyl-2,5-dimethylbenzene, 2-chloromethyl-p-xylene, benzene, 2-chloromethyl-1,4-dimethyl, 2-chloromethyl-1,4-dimethyl-benzene, benzenemethanol,2-bromo-5-fluoro, 2,5-dimethylbenzylchloride, acmc-209poz, 2.5-dimethylbenzylchloride
PubChem CID 69987
IUPAC Name 2-(chloromethyl)-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)CCl

Spécifications

Density 1.05
Boiling Point 232°C to 233°C
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.537
Quantity 25 g
UN Number UN3265
Beilstein 2206277
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylbenzyl chloride
Afficher plus de résultats Afficher moins de résultats
Hazard Category H227-H314
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 212-529-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only