missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzoxazole 98.0+%, TCI America™

Catalog No. p-7127740
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34615G 5 g
D346125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34615G Supplier TCI America Supplier No. D34615G
Only null left

Chemical Identifiers

CAS 5676-58-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00005773
InChI Key XVQGFGKAPKEUFT-UHFFFAOYSA-N
Synonym 2,5-dimethylbenzoxazole, 2,5-dimethylbenzo d oxazole, benzoxazole, 2,5-dimethyl, 2,5-dimethyl benzoxazole, pubchem8640, acmc-209lua, benzoxazole,5-dimethyl, 2,5-dimethyl-benzoxazole, benzoxazole,2,5-dimethyl, #
PubChem CID 21884
IUPAC Name 2,5-dimethyl-1,3-benzoxazole
SMILES CC1=CC2=C(C=C1)OC(=N2)C

Specifications

Color Yellow
Boiling Point 215°C
Quantity 5 g
Formula Weight 147.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylbenzoxazole
EINECSNumber (5)-0157
RTECSNumber DM4821600
TSCA Yes