missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzenethiol 98.0+%, TCI America™

Catalog No. D23935G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23935G Supplier TCI America Supplier No. D23935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4001-61-0
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00010020
InChI Key NHAUBUMQRJWWAT-UHFFFAOYSA-N
Synonym 2,5-dimethylthiophenol, 2,5-dimethyl thiophenol, benzenethiol, 2,5-dimethyl, 2,5-dimethylbenzene-1-thiol, 2,5-dimethylthiophenol;2,5-dimethylthiophenol;, p-xylenethiol, 2-mercapto-p-xylene, 2,5dimethylthiophenol, pubchem6811, 2,5-dimethyl-thiophenol
PubChem CID 77615
IUPAC Name 2,5-dimethylbenzenethiol
SMILES CC1=CC(=C(C=C1)C)S

Specifications

Color Colorless
Quantity 5 g
Formula Weight 138.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylbenzenethiol
EINECSNumber (3)-1094
TSCA Yes