missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzenesulfonic acid hydrate, 98%

CAS: 609-54-1 | C8H10O3S | 186.23 g/mol

Supplier:  Thermo Scientific Chemicals A1500722

 View more versions of this product

Catalog No. AAA1500722


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,5-Dimethylbenzenesulfonic acid hydrate
609-54-1
C8H10O3S
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
CC1=CC=C(C)C(=C1)S(O)(=O)=O
186.23
186.23 (Anhydrous)
84°C to 86°C
100 g
MFCD00151010
IRLYGRLEBKCYPY-UHFFFAOYSA-N
2,5-dimethylbenzenesulfonic acid
11868
99%

Chemical Identifiers

609-54-1
186.23
IRLYGRLEBKCYPY-UHFFFAOYSA-N
11868
CC1=CC=C(C)C(=C1)S(O)(=O)=O
C8H10O3S
MFCD00151010
p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
2,5-dimethylbenzenesulfonic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 210-193-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only