missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylbenzenesulfonic acid hydrate, 98%

Catalog No. AAA1500714
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1500714 25 g
AAA1500722 100 g
AAA1500736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1500714 Supplier Thermo Scientific Chemicals Supplier No. A1500714

CAS: 609-54-1 | C8H10O3S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 609-54-1
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00151010
InChI Key IRLYGRLEBKCYPY-UHFFFAOYSA-N
Synonym p-xylene-2-sulfonic acid, unii-l1sea5qnp9, p-xylene-2-sulphonic acid, benzenesulfonic acid, 2,5-dimethyl, l1sea5qnp9, 2,4-xylenesulfonic acid hydrate, 2,5-dimethyl-benzenesulfonic acid, 2,5-xylenesulfonic acid, p-xylenesulfonic acid, dsstox_cid_21641
PubChem CID 11868
IUPAC Name 2,5-dimethylbenzenesulfonic acid
SMILES CC1=CC=C(C)C(=C1)S(O)(=O)=O

Specifications

Melting Point 84°C to 86°C
Quantity 25 g
Formula Weight 186.23 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 2,5-Dimethylbenzenesulfonic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-193-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only