missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylanisole 98.0+%, TCI America™

Catalog No. D244025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D244025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D244025G Supplier TCI America Supplier No. D244025G
Only null left

Chemical Identifiers

CAS 1706-11-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008378
InChI Key SJZAUIVYZWPNAS-UHFFFAOYSA-N
Synonym 2,5-dimethylanisole, benzene, 2-methoxy-1,4-dimethyl, unii-rh47bom48g, 2-methoxy-p-xylene, rh47bom48g, 1,4-dimethyl-2-methyoxybenzene, benzene, 1-methoxy-2,5-dimethyl, 1,4-dimethyl-2-methoxybenzene, 2,5-dimethylanisol, pubchem5360
PubChem CID 74350
IUPAC Name 2-methoxy-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)OC

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 194°C
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,5-Dimethylanisole