missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylaniline 97.0+%, TCI America™

Catalog No. D066825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D066825G 25 g
D0668500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D066825G Supplier TCI America Supplier No. D066825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 95-78-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007743
InChI Key VOWZNBNDMFLQGM-UHFFFAOYSA-N
Synonym 2,5-xylidine, p-xylidine, 2,5-dimethylphenylamine, 2-amino-1,4-xylene, 2,5-dimethylbenzenamine, benzenamine, 2,5-dimethyl, 5-methyl-o-toluidine, 6-methyl-m-toluidine, 1-amino-2,5-dimethylbenzene, p-dimethylaniline
PubChem CID 7259
ChEBI CHEBI:518305
IUPAC Name 2,5-dimethylaniline
SMILES CC1=CC=C(C)C(N)=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 1711
Boiling Point 104°C
Formula Weight 121.18
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 2,5-Dimethylaniline
EINECSNumber (3)-0129
RTECSNumber ZE9100000
TSCA Yes