missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethylacetophenone, 97%

Catalog No. p-7081315
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1143206 5 g
AAA1143214 25 g
AAA1143222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1143206 Supplier Thermo Scientific Chemicals Supplier No. A1143206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2142-73-6 | C10H12O | 148.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2142-73-6
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00014988
InChI Key AWKBVLVKQQRRFQ-UHFFFAOYSA-N
Synonym 2',5'-dimethylacetophenone, 2,5-dimethylacetophenone, 1-2,5-dimethylphenyl ethanone, 1-2,5-dimethylphenyl ethan-1-one, ethanone, 1-2,5-dimethylphenyl, acetophenone, 2',5'-dimethyl, 1-acetyl-2,5-dimethylbenzene, 2-acetyl-p-xylene, acmc-1cliy, 2',5'dimethylacetophenone
PubChem CID 75061
IUPAC Name 1-(2,5-dimethylphenyl)ethanone
SMILES CC1=CC(=C(C=C1)C)C(=O)C

Specifications

Density 1
Boiling Point 230°C to 231°C
Flash Point 95°C (203°F)
Odor Characteristic
Refractive Index 1.529
Quantity 5 g
Beilstein 2042139
Formula Weight 148.21
Percent Purity ≥98%
Chemical Name or Material 2',5'-Dimethylacetophenone
Hazard Category H302+H332-H319
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
EINECSNumber 218-399-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only