missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™

$111.51 - $361.67

Chemical Identifiers

CAS 59480-92-1
Molecular Formula C6H12N
Molecular Weight (g/mol) 98.17
MDL Number MFCD00005215
InChI Key TXQDHQBSNAJSHQ-OLQVQODUSA-O
PubChem CID 101066
IUPAC Name 2,5-dimethyl-2,5-dihydro-1H-pyrrole
SMILES C[C@H]1[NH2+][C@@H](C)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC165930010
SDS View Documents
Thermo Scientific Chemicals
165930010
1 g Glass bottle
Each for $111.51
Only null left
 
AC165930050
SDS View Documents
Thermo Scientific Chemicals
165930050
5 g Glass bottle
Each for $361.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

59480-92-1
98.17
TXQDHQBSNAJSHQ-OLQVQODUSA-O
2,5-dimethyl-2,5-dihydro-1H-pyrrole
C6H12N
MFCD00005215
101066
C[C@H]1[NH2+][C@@H](C)C=C1

Specifications

59480-92-1
100.0
0.8240g/mL
Authentic
Glass bottle
1.4404
1 g
2,5-dimethyl-3-pyrroline, 2,5-dimethyl-3-pyrroline, mixture of cis and trans, 2,5-dimethyl-2,5-dihydropyrrole, 1h-pyrrole,2,5-dihydro-2,5-dimethyl, 1h-pyrrole, 2,5-dihydro-2,5-dimethyl
C[C@H]1[NH2+][C@@H](C)C=C1
98.17
97.16
Liquid
70.0
Colorless to Yellow
5°C
70% min. (GC)
C6H12N
MFCD00005215
0.824
TXQDHQBSNAJSHQ-OLQVQODUSA-O
2,5-dimethyl-2,5-dihydro-1H-pyrrole
101066
75%
2, 5-Dimethyl-3-pyrroline, 75%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 261-781-3

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only