missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-pyrroline, 75%, mixture of cis and trans, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 165930050

 View more versions of this product

Catalog No. AC165930050

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2, 5-Dimethyl-3-pyrroline
59480-92-1
100.0
0.8240g/mL
Authentic
Glass bottle
1.4404
5 g
2,5-dimethyl-3-pyrroline, 2,5-dimethyl-3-pyrroline, mixture of cis and trans, 2,5-dimethyl-2,5-dihydropyrrole, 1h-pyrrole,2,5-dihydro-2,5-dimethyl, 1h-pyrrole, 2,5-dihydro-2,5-dimethyl
C[C@H]1[NH2+][C@@H](C)C=C1
98.17
97.16
Liquid
75%
70.0
Colorless to Yellow
5°C
70% min. (GC)
C6H12N
MFCD00005215
0.824
TXQDHQBSNAJSHQ-OLQVQODUSA-O
2,5-dimethyl-2,5-dihydro-1H-pyrrole
101066
75%

Chemical Identifiers

59480-92-1
98.17
TXQDHQBSNAJSHQ-OLQVQODUSA-O
2,5-dimethyl-2,5-dihydro-1H-pyrrole
C6H12N
MFCD00005215
101066
C[C@H]1[NH2+][C@@H](C)C=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 261-781-3

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only