missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3-hexyne-2,5-diol 99.0+%, TCI America™

Catalog No. p-7126407
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
D073825G 25 g
D0738500G 500 g
2 options

Catalog No. D073825G

Supplier: TCI America D073825G

Only null left

Chemical Identifiers

CAS 142-30-3
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.198
MDL Number MFCD00004468
InChI Key IHJUECRFYCQBMW-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol
PubChem CID 8883
IUPAC Name 2,5-dimethylhex-3-yne-2,5-diol
SMILES CC(C)(C#CC(C)(C)O)O

Specifications

Melting Point 95°C
Color White-Yellow
Boiling Point 206°C
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethyl-3-hexyne-2,5-diol
EINECSNumber (2)-0270
RTECSNumber MR0176000
TSCA Yes