accessibility menu, dialog, popup

UserName

CH3CH3OHH3CCH3OH

CAS RN 142-30-3

IUPAC Name: 2,5-dimethylhex-3-yne-2,5-diol
Synonyms: 2,5-dimethyl-3-hexyne-2,5-diol
Molecular Weight (g/mol): 142.2
Molecular Formula: C8H14O2
InChi Key: IHJUECRFYCQBMW-UHFFFAOYSA-N
SMILES: CC(C)(O)C#CC(C)(C)O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)