missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-2-hexene 96.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 113°C |
| Quantity | 10 mL |
| UN Number | 3295 |
| Formula Weight | 112.22 |
| Percent Purity | ≥96.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,5-Dimethyl-2-hexene |
Safety and Handling
| TSCA | No |