accessibility menu, dialog, popup

UserName

CH3CH3H3CH3C

CAS RN 3404-78-2

IUPAC Name: 2,5-dimethylhex-2-ene
Synonyms: 2,5-dimethyl-2-hexene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: VFZIUYUUQFYZBR-UHFFFAOYSA-N
SMILES: CC(C)CC=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results