missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-2,4-hexadiene (stabilized with BHT) 96.0+%, TCI America™
$52.19 - $346.71
Chemical Identifiers
| CAS | 764-13-6 |
|---|---|
| Molecular Formula | C8H14 |
| Molecular Weight (g/mol) | 110.20 |
| MDL Number | MFCD00008904 |
| InChI Key | DZPCYXCBXGQBRN-UHFFFAOYSA-N |
| Synonym | 2,5-dimethyl-2,4-hexadiene, biisobutenyl, biisocrotyl, diisocrotyl, 2,4-hexadiene, 2,5-dimethyl, unii-ee5yri32x6, ee5yri32x6, 2,4-diene, acmc-1bi1p, 2,5dimethyl-2,4-hexadiene |
| PubChem CID | 12992 |
| IUPAC Name | 2,5-dimethylhexa-2,4-diene |
| SMILES | CC(C)=CC=C(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D073525ML
|
TCI America
D073525ML |
25 mL |
Each for $52.19
|
|
|||||
|
D0735500ML
|
TCI America
D0735500ML |
500 mL |
Each for $346.71
|
|
|||||
Chemical Identifiers
| 764-13-6 | |
| 110.20 | |
| DZPCYXCBXGQBRN-UHFFFAOYSA-N | |
| 12992 | |
| CC(C)=CC=C(C)C |
| C8H14 | |
| MFCD00008904 | |
| 2,5-dimethyl-2,4-hexadiene, biisobutenyl, biisocrotyl, diisocrotyl, 2,4-hexadiene, 2,5-dimethyl, unii-ee5yri32x6, ee5yri32x6, 2,4-diene, acmc-1bi1p, 2,5dimethyl-2,4-hexadiene | |
| 2,5-dimethylhexa-2,4-diene |
Specifications
| 764-13-6 | |
| 135°C | |
| MFCD00008904 | |
| 3295 | |
| DZPCYXCBXGQBRN-UHFFFAOYSA-N | |
| 2,5-dimethylhexa-2,4-diene | |
| 12992 | |
| ≥96.0% (GC) | |
| 2,5-Dimethyl-2,4-hexadiene (stabilized with BHT) |
| Yellow | |
| C8H14 | |
| 25 mL | |
| 2,5-dimethyl-2,4-hexadiene, biisobutenyl, biisocrotyl, diisocrotyl, 2,4-hexadiene, 2,5-dimethyl, unii-ee5yri32x6, ee5yri32x6, 2,4-diene, acmc-1bi1p, 2,5dimethyl-2,4-hexadiene | |
| CC(C)=CC=C(C)C | |
| 110.20 | |
| 110.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0028
TSCA : Yes