accessibility menu, dialog, popup

UserName

CAS RN 764-13-6

CH3CH3H3CH3C

CAS RN 764-13-6

IUPAC Name: 2,5-dimethylhexa-2,4-diene
Synonyms: 2,5-dimethyl-2,4-hexadiene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: DZPCYXCBXGQBRN-UHFFFAOYSA-N
SMILES: CC(C)=CC=C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results