missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde, 98+%

Catalog No. AAL0583606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0583606 5 g
AAL0583614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0583606 Supplier Thermo Scientific Chemicals Supplier No. L0583606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 83-18-1 | C13H13NO | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83-18-1
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.25
MDL Number MFCD00051494
InChI Key LNROIXNEIZSESG-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1-phenyl-1h-pyrrole-3-carbaldehyde, 2,5-dimethyl-1-phenylpyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxaldehyde, 2,5-dimethyl-1-phenyl, 2,5-dimethyl-1-phenyl-1h-pyrrole-3-carboxaldehyde, osm-s-28, 10.14272/lnroixneizsesg-uhfffaoysa-n, 2,5-dimethyl-1-phenyl-3-pyrrolecarboxaldehyde, pyrrole-3-carboxaldehyde,5-dimethyl-1-phenyl, 2,5-dimethyl-1-phenyl pyrrole-3-carboxaldehyde, 1-phenyl-2,5-dimethyl-1h-pyrrole-3-carbaldehyde
PubChem CID 66515
IUPAC Name 2,5-dimethyl-1-phenylpyrrole-3-carbaldehyde
SMILES CC1=CC(C=O)=C(C)N1C1=CC=CC=C1

Specifications

Melting Point 87°C to 90°C
Quantity 5 g
Beilstein 144790
Sensitivity Air sensitive
Formula Weight 199.25
Percent Purity ≥98%
Chemical Name or Material 2,5-Dimethyl-1-phenylpyrrole-3-carboxaldehyde
EINECSNumber 201-458-6
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only