accessibility menu, dialog, popup

UserName

CAS RN 83-18-1

IUPAC Name: 2,5-dimethyl-1-phenyl-1H-pyrrole-3-carbaldehyde
Synonyms: 2,5-dimethyl-1-phenylpyrrole-3-carbaldehyde
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: LNROIXNEIZSESG-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=C(C)N1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results