missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,4-phenylenediamine 98.0+%, TCI America™

Supplier:  TCI America D2183100G

 View more versions of this product

Catalog No. D2183100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,5-Dimethyl-1,4-phenylenediamine
6393-01-7
Crystal-Powder at 20°C
MFCD00052675
BWAPJIHJXDYDPW-UHFFFAOYSA-N
2,5-dimethylbenzene-1,4-diamine
160824
136.20
148°C
100 g
C8H12N2
2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4
CC1=CC(=C(C=C1N)C)N
136.20
CHEBI:53613
≥98.0% (GC,T)

Chemical Identifiers

6393-01-7
136.20
BWAPJIHJXDYDPW-UHFFFAOYSA-N
160824
2,5-dimethylbenzene-1,4-diamine
C8H12N2
MFCD00052675
2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4
CHEBI:53613
CC1=CC(=C(C=C1N)C)N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions