missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-1,4-phenylenediamine 98.0+%, TCI America™

Catalog No. D218325G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D218325G 25 g
D2183100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D218325G Supplier TCI America Supplier No. D218325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6393-01-7 | C8H12N2 | 136.20 g/mol

Chemical Identifiers

CAS 6393-01-7
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00052675
InChI Key BWAPJIHJXDYDPW-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4
PubChem CID 160824
ChEBI CHEBI:53613
IUPAC Name 2,5-dimethylbenzene-1,4-diamine
SMILES CC1=CC(=C(C=C1N)C)N

Specifications

Melting Point 148°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 136.20
Percent Purity ≥98.0% (GC,T)
Grade Not Available
Chemical Name or Material 2,5-Dimethyl-1,4-phenylenediamine