missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxytoluene 98.0+%, TCI America™

Catalog No. D25525G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25525G 5 g
D255225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25525G Supplier TCI America Supplier No. D25525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24599-58-4
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00041901
InChI Key IQISOVKPFBLQIQ-UHFFFAOYSA-N
Synonym 2,5-dimethoxytoluene, benzene, 1,4-dimethoxy-2-methyl, toluene, 2,5-dimethoxy, methyl hydroquinone dimethyl ester, 2-methylhydroquinone dimethyl ether, 1,4-dimethoxy-2-methyl-benzene, pubchem4543, 2,5-dimethoxy toluene, acmc-209gdc
PubChem CID 90552
IUPAC Name 1,4-dimethoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)OC

Specifications

Color Yellow
Boiling Point 218°C
Quantity 5 g
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethoxytoluene