missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzyl alcohol, 99%

Catalog No. p-114335
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC182200050 5 mL Glass bottle
AC182200250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC182200050 Supplier Thermo Scientific Chemicals Supplier No. 182200050

CAS: 33524-31-1 | C9H12O3 | 168.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33524-31-1
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004615
InChI Key WGQMUABRZGUAOS-UHFFFAOYSA-N
Synonym 2,5-dimethoxybenzyl alcohol, 2,5-dimethoxyphenyl methanol, benzenemethanol, 2,5-dimethoxy, acmc-1ckmt, 2,5-dimethoxybenzylalcohol, 2,5-dimethoxylbenzylalcohol, benzenemethanol,2,5-dimethoxy, 2,5-dimethoxyphenyl-methanol, wgqmuabrzguaos-uhfffaoysa, 2,5-dimethoxyphenyl methanol #
PubChem CID 118495
IUPAC Name (2,5-dimethoxyphenyl)methanol
SMILES COC1=CC(=C(C=C1)OC)CO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 122°C to 125°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5460 to 1.548
Linear Formula (CH3O)2C6H3CH2OH
Quantity 5 mL
Formula Weight 168.19
Percent Purity 99%
Physical Form Viscous Liquid
Chemical Name or Material 2, 5-Dimethoxybenzyl alcohol, 99%
Show More Show Less
EINECSNumber 251-562-0
TSCA TSCA

RUO – Research Use Only