missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxybenzonitrile 98.0+%, TCI America™

Catalog No. D36591G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D36591G 1 g
D36595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D36591G Supplier TCI America Supplier No. D36591G

Chemical Identifiers

CAS 5312-97-0
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00016375
InChI Key HWAMEJIMXIXLIH-UHFFFAOYSA-N
Synonym benzonitrile, 2,5-dimethoxy, 2,5-dimethoxybenzenecarbonitrile, pubchem13109, acmc-1an8e, benzonitrile,2,5-dimethoxy, 2-cyano-1,4-dimethoxybenzene, 2,5-dimethoxybenzonitrile, labotest-bb lt00848082, 2-cyanohydroquinone dimethyl ether
PubChem CID 79200
IUPAC Name 2,5-dimethoxybenzonitrile
SMILES COC1=CC(=C(C=C1)OC)C#N

Specifications

Melting Point 83°C
Quantity 1 g
UN Number 3439
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxybenzonitrile