missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyaniline 98.0+%, TCI America™

Catalog No. p-7126997
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D189325G 25 g
D1893500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D189325G Supplier TCI America Supplier No. D189325G
Only null left

Chemical Identifiers

CAS 102-56-7
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008368
InChI Key NAZDVUBIEPVUKE-UHFFFAOYSA-N
Synonym benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline
PubChem CID 7613
IUPAC Name 2,5-dimethoxyaniline
SMILES COC1=CC=C(OC)C(N)=C1

Specifications

Melting Point 81°C
Boiling Point 159°C
Quantity 25 g
UN Number 2811
Formula Weight 153.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethoxyaniline
EINECSNumber (3)-0616
RTECSNumber BX4375000
TSCA Yes