missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyaniline, 99%

Catalog No. p-7044181
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0505330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0505330 Supplier Thermo Scientific Chemicals Supplier No. L0505330
Only null left

CAS: 102-56-7 | C8H11NO2 | 153.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 102-56-7
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008368
InChI Key NAZDVUBIEPVUKE-UHFFFAOYSA-N
Synonym benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline
PubChem CID 7613
IUPAC Name 2,5-dimethoxyaniline
SMILES COC1=CC=C(OC)C(N)=C1

Specifications

Melting Point 78°C to 82°C
Boiling Point 140°C (7 mmHg)
Flash Point 130°C (266°F)
Odor Characteristic
Quantity 250 g
UN Number UN2811
Beilstein 776823
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2,5-Dimethoxyaniline
Hazard Category H301+H311+H331-H373
Hazard Statement GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-040-9
RTECSNumber BX4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only