missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethoxyacetanilide 98.0+%, TCI America™

Catalog No. D167125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D167125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D167125G Supplier TCI America Supplier No. D167125G
Only null left

Chemical Identifiers

CAS 3467-59-2
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00043826
InChI Key HOZRMSVNIKYCMB-UHFFFAOYSA-N
Synonym 2-Acetamido-1,4-dimethoxybenzene
PubChem CID 77015
IUPAC Name N-(2,5-dimethoxyphenyl)acetamide
SMILES COC1=CC=C(OC)C(NC(C)=O)=C1

Specifications

Melting Point 91°C
Quantity 25 g
Formula Weight 195.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,5′-Dimethoxyacetanilide