missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxy-4-methylbenzaldehyde, 97%

Catalog No. AAH6414003
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6414003 1 g
AAH6414006 5 g
AAH6414014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6414003 Supplier Thermo Scientific Chemicals Supplier No. H6414003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4925-88-6 | C10H12O3 | 180.20 g/mol

Used in the preparation of phenylamine derivative as potential psychotomimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of phenylamine derivative as potential psychotomimetics.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 4925-88-6
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD02253192
InChI Key LRSRTWLEJBIAIT-UHFFFAOYSA-N
Synonym 4-methyl-2,5-dimethoxybenzaldehyde, unii-238b3l4auk, 2,5-dimethoxy-4-methyl-benzaldehyde, benzaldehyde, 2,5-dimethoxy-4-methyl, 2,5-dimethoxy-p-tolualdehyde, p-tolualdehyde, 2,5-dimethoxy, who-dd, 2,5-dimethoxy-4-methylbenzaldehyde;, 2,5-dimethoxy-4-methyl-benzaldehyde;, #
PubChem CID 602019
IUPAC Name 2,5-dimethoxy-4-methylbenzaldehyde
SMILES COC1=CC(C)=C(OC)C=C1C=O

Specifications

Melting Point 82°C to 86°C
Color White
Quantity 1 g
Beilstein 1866592
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 180.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2,5-Dimethoxy-4-methylbenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only