accessibility menu, dialog, popup

UserName

CAS RN 4925-88-6

IUPAC Name: 2,5-dimethoxy-4-methylbenzaldehyde
Synonyms: 2,5-dimethoxy-4-methylbenzaldehyde
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: LRSRTWLEJBIAIT-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(OC)C=C1C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results