missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethoxy-2,5-dihydrofuran (cis- and trans- mixture) 98.0+%, TCI America™
Supplier: TCI America D175925G
Specifications
2,5-Dimethoxy-2,5-dihydrofuran (cis- and trans- mixture) | |
Yellow | |
MFCD00003220 | |
1993 | |
WXFWXFIWDGJRSC-UHFFFAOYNA-N | |
2,5-dimethoxy-2,5-dihydrofuran | |
78974 | |
≥98.0% (GC) |
332-77-4 | |
C6H10O3 | |
25 g | |
2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans | |
COC1OC(OC)C=C1 | |
130.14 | |
130.14 | |
Liquid |
Chemical Identifiers
332-77-4 | |
130.14 | |
WXFWXFIWDGJRSC-UHFFFAOYNA-N | |
78974 | |
COC1OC(OC)C=C1 |
C6H10O3 | |
MFCD00003220 | |
2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + transShow More | |
2,5-dimethoxy-2,5-dihydrofuran |
Safety and Handling
TSCA : Yes