missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diiodothiophene 97.0+%, TCI America™

Catalog No. D26115G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26115G Supplier TCI America Supplier No. D26115G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 625-88-7
Molecular Formula C4H2I2S
Molecular Weight (g/mol) 335.929
MDL Number MFCD00014525
InChI Key PNYWRAHWEIOAGK-UHFFFAOYSA-N
Synonym thiophene, 2,5-diiodo, 2,5-diiodthiophen, thiophene,5-diiodo, pubchem5529, maybridge1_002904, acmc-1b0lo, 2,5-diiodothiophene, bidd:gt0169, thiophene, 2,5-diiodo-8ci 9ci
PubChem CID 69368
IUPAC Name 2,5-diiodothiophene
SMILES C1=C(SC(=C1)I)I

Specifications

Melting Point 41°C
Boiling Point 140°C
Quantity 5 g
Formula Weight 335.93
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diiodothiophene
TSCA No
Recommended Storage Refrigerator