accessibility menu, dialog, popup

UserName

ISI

CAS RN 625-88-7

IUPAC Name: 2,5-diiodothiophene
Synonyms: 2,5-diiodothiophene
Molecular Weight (g/mol): 335.93
Molecular Formula: C4H2I2S
InChi Key: PNYWRAHWEIOAGK-UHFFFAOYSA-N
SMILES: IC1=CC=C(I)S1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)