missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxyphenylacetic acid, 97%, Thermo Scientific™

Catalog No. AAH60742ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60742ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH60742ME Supplier Thermo Scientific Chemicals Supplier No. H60742ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an intermediate in the metabolism of tyrosine and phenylalanine.

Solubility
Soluble in methanol.

Notes
Store at cool temperature.

Chemical Identifiers

CAS 451-13-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00004324
InChI Key IGMNYECMUMZDDF-UHFFFAOYSA-N
Synonym homogentisic acid, 2,5-dihydroxyphenylacetic acid, alcapton, homogentisate acid, homogentisinic acid, 2,5-dihydroxyphenyl acetic acid, homogentisate, benzeneacetic acid, 2,5-dihydroxy, 2-2,5-dihydroxyphenyl acetic acid, 2,5-dihydroxy-alpha-toluic acid
PubChem CID 780
ChEBI CHEBI:44747
IUPAC Name 2-(2,5-dihydroxyphenyl)acetic acid
SMILES OC(=O)CC1=CC(O)=CC=C1O

Specifications

Melting Point 150°C to 152°C
Quantity 500 mg
Beilstein 2692860
Solubility Information Soluble in methanol.
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material 2,5-Dihydroxyphenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-192-7
RTECSNumber AH0589000
TSCA Yes
Recommended Storage Ambient temperatures