missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydroxybenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 206°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 154.12 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,5-Dihydroxybenzoic Acid |
Safety and Handling
| EINECSNumber | (3)-1616&(3)-3065 |
| RTECSNumber | LY3850000 |
| TSCA | Yes |