missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid 99.0+%, TCI America™

Catalog No. p-7127570
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
D29331G 1 g
D29335G 5 g
2 options

Catalog No. D29331G

Supplier: TCI America D29331G

Only null left

Chemical Identifiers

CAS 490-79-9
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002460
InChI Key WXTMDXOMEHJXQO-UHFFFAOYSA-N
Synonym gentisic acid, hydroquinonecarboxylic acid, benzoic acid, 2,5-dihydroxy, 5-hydroxysalicylic acid, gensigen, gensigon, gentisate, 2,5-dioxybenzoic acid, 2,5-dhba, gentisinic acid
PubChem CID 3469
ChEBI CHEBI:17189
IUPAC Name 2,5-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)O

Specifications

Melting Point 206°C
Color White-Yellow
Quantity 1 g
Formula Weight 154.12
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dihydroxybenzoic Acid [Matrix for MALDI-TOF/MS]
EINECSNumber (3)-1616&(3)-3065
RTECSNumber LY3850000
TSCA Yes