missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic acid, 99%

Catalog No. AAA1145914
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1145914 25 g
AAA1145922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1145914 Supplier Thermo Scientific Chemicals Supplier No. A1145914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 490-79-9 | C7H6O4 | 154.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2,5-dihydroxybenzoic acid are used as intermediates for pharmaceuticals (especially for antipyetic anlgesic, antirheumatism) and other organic synthesis. They are used as matrix for ionization of peptides, proteins and carbohydrates. In industrial field, they are used as intermediates for the production of other organic chemicals, resins, polyesters, plasticizers, dyestuffs, preservatives, and rubber chemicals.

Solubility
It is soluble in alcohol and ether.

Notes
Keep away from Strong oxidizing agents, Strong bases, Acid chlorides, Acid anhydrides. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 490-79-9
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002460
InChI Key WXTMDXOMEHJXQO-UHFFFAOYSA-N
Synonym gentisic acid, hydroquinonecarboxylic acid, benzoic acid, 2,5-dihydroxy, 5-hydroxysalicylic acid, gensigen, gensigon, gentisate, 2,5-dioxybenzoic acid, 2,5-dhba, gentisinic acid
PubChem CID 3469
ChEBI CHEBI:17189
IUPAC Name 2,5-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)O

Specifications

Melting Point ∼206°C (decomposition)
Quantity 25 g
Beilstein 2209119
Merck Index 14,4398
Solubility Information It is soluble in alcohol and ether.
Formula Weight 154.12
Percent Purity 99%
Chemical Name or Material 2,5-Dihydroxybenzoic acid
Hazard Category H302-H315-H319-H335-H500
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-718-5
RTECSNumber LY3850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only