Learn More
2,5-Dihydroxybenzoic acid, 99%, Thermo Scientific™
$26.38 - $26.38
Chemical Identifiers
CAS | 490-79-9 |
---|---|
Molecular Formula | C7H6O4 |
Molecular Weight (g/mol) | 154.12 |
MDL Number | MFCD00002460 |
InChI Key | WXTMDXOMEHJXQO-UHFFFAOYSA-N |
Synonym | 2,5-dhba, 2,5-dioxybenzoic acid, 5-hydroxysalicylic acid, benzoic acid, 2,5-dihydroxy, gensigen, gensigon, gentisate, gentisic acid, gentisinic acid, hydroquinonecarboxylic acid |
PubChem CID | 3469 |
ChEBI | CHEBI:17189 |
IUPAC Name | 2,5-dihydroxybenzoic acid |
SMILES | C1=CC(=C(C=C1O)C(=O)O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC165200050
|
Fisher Scientific Acros
165200050 |
5g | Glass bottle |
Each for $26.38
|
|
AC165200500
|
Fisher Scientific Acros
165200500 |
50g | Plastic bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC165202500
|
Thermo Fisher Scientific
AC165202500 |
250g | Plastic bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
490-79-9 | |
154.12 | |
WXTMDXOMEHJXQO-UHFFFAOYSA-N | |
3469 | |
2,5-dihydroxybenzoic acid |
C7H6O4 | |
MFCD00002460 | |
2,5-dhba, 2,5-dioxybenzoic acid, 5-hydroxysalicylic acid, benzoic acid, 2,5-dihydroxy, gensigen, gensigon, gentisate, gentisic acid, gentisinic acid, hydroquinonecarboxylic acid | |
CHEBI:17189 | |
C1=CC(=C(C=C1O)C(=O)O)O |
Specifications
490-79-9 | |
100.0 | |
204.0°C to 207.0°C | |
Authentic | |
(HO)2C6H3CO2H | |
10, 384 | |
15, 4433 | |
5g | |
WXTMDXOMEHJXQO-UHFFFAOYSA-N | |
2,5-dihydroxybenzoic acid | |
3469 | |
154.12 | |
99% |
98.5 | |
White to Beige | |
99% | |
C7H6O4 | |
MFCD00002460 | |
Glass bottle | |
2,5-dhba, 2,5-dioxybenzoic acid, 5-hydroxysalicylic acid, benzoic acid, 2,5-dihydroxy, gensigen, gensigon, gentisate, gentisic acid, gentisinic acid, hydroquinonecarboxylic acid | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ether,practically insoluble in chloroform,benzene and,carbon disulfide | |
C1=CC(=C(C=C1O)C(=O)O)O | |
154.12 | |
CHEBI:17189 | |
Crystalline Powder | |
2, 5-Dihydroxybenzoic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
GHS Signal Word: Warning
EINECSNumber : 207-718-5
RUO – Research Use Only