missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydroxybenzaldehyde 98.0+%, TCI America™
$51.98 - $180.97
Chemical Identifiers
| CAS | 1194-98-5 |
|---|---|
| Molecular Formula | C7H6O3 |
| Molecular Weight (g/mol) | 138.12 |
| MDL Number | MFCD00003333 |
| InChI Key | CLFRCXCBWIQVRN-UHFFFAOYSA-N |
| Synonym | gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy |
| PubChem CID | 70949 |
| ChEBI | CHEBI:28508 |
| IUPAC Name | 2,5-dihydroxybenzaldehyde |
| SMILES | OC1=CC=C(O)C(C=O)=C1 |
Chemical Identifiers
| 1194-98-5 | |
| 138.12 | |
| CLFRCXCBWIQVRN-UHFFFAOYSA-N | |
| 70949 | |
| 2,5-dihydroxybenzaldehyde |
| C7H6O3 | |
| MFCD00003333 | |
| gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy | |
| CHEBI:28508 | |
| OC1=CC=C(O)C(C=O)=C1 |
Specifications
| 1194-98-5 | |
| C7H6O3 | |
| 1 g | |
| CLFRCXCBWIQVRN-UHFFFAOYSA-N | |
| 2,5-dihydroxybenzaldehyde | |
| 70949 | |
| 138.12 | |
| Crystalline Powder |
| 102°C | |
| MFCD00003333 | |
| gentisaldehyde, benzaldehyde, 2,5-dihydroxy, gentisate aldehyde, unii-0q83hds90w, polymer, 2-formylhydroquinone, pubchem2285, acmc-1btnd, 2,5 dihydroxybenzaldehyde, benzaldehyde,5-dihydroxy | |
| OC1=CC=C(O)C(C=O)=C1 | |
| 138.12 | |
| CHEBI:28508 | |
| ≥98.0% (GC) | |
| 2,5-Dihydroxybenzaldehyde |
Safety and Handling
TSCA : Yes