accessibility menu, dialog, popup

UserName

CAS RN 1194-98-5

IUPAC Name: 2,5-dihydroxybenzaldehyde
Synonyms: 2,5-dihydroxybenzaldehyde gentisaldehyde gentisate aldehyde
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: CLFRCXCBWIQVRN-UHFFFAOYSA-N
SMILES: OC1=CC=C(O)C(C=O)=C1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)